Learn More
Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade
A lipotropic agent
74.97 € - 262.31 €
Chemical Identifiers
CAS | 87-89-8 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
MDL Number | MFCD00077932 |
InChI Key | CDAISMWEOUEBRE-UHFFFAOYSA-N |
Synonym | scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol |
PubChem CID | 892 |
ChEBI | CHEBI:24848 |
IUPAC Name | cyclohexane-1,2,3,4,5,6-hexol |
SMILES | C1(C(C(C(C(C1O)O)O)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15495179
|
Thermo Scientific Chemicals
J62886.18 |
50 g |
74.97 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405189
|
Thermo Scientific Chemicals
J62886.22 |
100 g |
131.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15415189
|
Thermo Scientific Chemicals
J62886.30 |
250 g |
262.31 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
Solubility
It is soluble in water (50 mg/ml).
Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
87-89-8 | |
180.156 | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
892 | |
cyclohexane-1,2,3,4,5,6-hexol |
C6H12O6 | |
MFCD00077932 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
CHEBI:24848 | |
C1(C(C(C(C(C1O)O)O)O)O)O |
Specifications
87-89-8 | |
White | |
C6H12O6 | |
1907329 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
cyclohexane-1,2,3,4,5,6-hexol | |
892 | |
180.16 | |
Crystalline Powder |
222°C to 227°C | |
Odorless | |
MFCD00077932 | |
14,4978 | |
It is soluble in water (50mg/ml). | |
C1(C(C(C(C(C1O)O)O)O)O)O | |
180.156 | |
CHEBI:24848 | |
Cell Culture | |
myo-Inositol |
Safety and Handling
missing translation for 'einecsNumber' : 201-781-2
missing translation for 'rtecsNumber' : NM7520800
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.