missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-1-Hexane sulfonic acid, Sodium Salt, Monhydrate, 99.3 %, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 2832-45-3 |
Form | Crystalline Powder |
Quantity | 10 g |
Appearance | White |
Molecular Formula | C6H13NaO3S |
MDL Number | MFCD00007542 |
Synonym | sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt |
InChI Key | QWSZRRAAFHGKCH-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCCS([O-])(=O)=O |
IUPAC Name | sodium hexane-1-sulfonate |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
n-1-Hexane sulfonic acid, Sodium Salt, Monhydrate, 99.3 %, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction