missing translation for 'onlineSavingsMsg'
Learn More

n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™

Used as an ion pairing reagent for HPLC and as an anionic surfactant. n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals has a low molecular weight that will not form micelles in solution.

Brand:  MP Biomedicals 0215125205

 View more versions of this product

Product Code. 17871371

  • 42.40 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.

  • The anionic sulfonate counterion permits the separation and resolution of positively charged analytes
  • Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value
  • For ion-pair chromatography
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2–8°C
C6H13NaO3S
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium hexane-1-sulfonate
5 g
MFCD00007542
[Na+].CCCCCCS([O-])(=O)=O
188.22
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™ > 5g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.