Learn More
N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
53.00 € - 180.00 €
Chemical Identifiers
CAS | 1465-25-4 |
---|---|
Molecular Formula | C12H16Cl2N2 |
Molecular Weight (g/mol) | 259.174 |
MDL Number | MFCD00012556 |
InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Synonym | n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride |
PubChem CID | 15106 |
ChEBI | CHEBI:53452 |
IUPAC Name | N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
SMILES | C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493377
|
Thermo Scientific Alfa Aesar
A17164.09 |
10 g |
53.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403387
|
Thermo Scientific Alfa Aesar
A17164.18 |
50 g |
180.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.
Solubility
Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.
Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
Chemical Identifiers
1465-25-4 | |
259.174 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
15106 | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
C12H16Cl2N2 | |
MFCD00012556 | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
CHEBI:53452 | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Specifications
1465-25-4 | |
C12H16Cl2N2 | |
3707471 | |
14,6409 | |
Soluble in hot water,alcohol,dilute hydrochloric acid and acetone. | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
259.174 | |
CHEBI:53452 | |
96% |
191°C to 199°C | |
MFCD00012556 | |
Hygroscopic; Light sensitive | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
15106 | |
259.18 | |
N-(1-Naphthyl)ethylenediamine dihydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-981-2
missing translation for 'rtecsNumber' : KV5330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- N-(1-Naphthyl)ethylenediamine dihydrochloride
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only