missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-1-Naphthylethylene Diamine Dihydrochloride, Extra Pure, SLR, Fisher Chemical™
84.45 € - 715.00 €
Chemical Identifiers
CAS | 1465-25-4 |
---|---|
Molecular Formula | C12H16Cl2N2 |
Molecular Weight (g/mol) | 259.174 |
MDL Number | 12556 |
InChI Key | MZNYWPRCVDMOJG-UHFFFAOYSA-N |
Synonym | n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride |
PubChem CID | 15106 |
ChEBI | CHEBI:53452 |
IUPAC Name | N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
SMILES | C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Description
N-1-Naphthylethylene Diamine Dihydrochloride, Extra Pure, SLR
Chemical Identifiers
1465-25-4 | |
259.174 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
15106 | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride |
C12H16Cl2N2 | |
12556 | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
CHEBI:53452 | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl |
Specifications
1465-25-4 | |
Beige | |
C12H16Cl2N2 | |
n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride | |
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl | |
259.174 | |
CHEBI:53452 | |
Solid |
200°C | |
Glass Jar | |
12556 | |
MZNYWPRCVDMOJG-UHFFFAOYSA-N | |
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride | |
15106 | |
259.17g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- N-(1-Naphthyl)ethylenediamine, dihydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/A§/if you feel unwell.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title