Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
Brand:  Macherey-Nagel™ 70231.3
 View more versions of this product
Product Code. 10652034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Ready assembled crimp closures and plain crimp caps N 11 
  • Septa are assembled, caps are ready-to-use.
  • Septa are shown for illustration purposes only, images upside down.
  • Packs of 100 closures each
Specifications
Specifications
Blue
Natural rubber / Butyl red orange/ TEF colorless
1 mm
Aluminum, Butyl Rubber, Rubber, TEF
100 Pk.
Crimp Cap