Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
14.40 € - 34.20 €
Product Code | Brand | Thickness (Metric) | Color | Septum | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (Metric) | Color | Septum | Price | Quantity & Availability | |||
10186563
|
Macherey-Nagel™
70284 |
0.25 mm | Silver | PTFE Virginal/White |
14.40 €
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10468852
|
Macherey-Nagel™
70288 |
1.3 mm | Silver | Silicone White/PTFE Red |
25.50 €
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12490500
|
Macherey-Nagel™
702995 |
1 mm | Silver | PTFE Red /Silicone White/PTFE Red |
30.90 €
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12420460
|
Macherey-Nagel™
702823 |
1.5 mm | Silver | Silicone White/PTFE Blue Cross-slit |
34.20 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||
10017630
|
Macherey-Nagel™
70231 |
1.3 mm | Silver | Natural rubber / Butyl red orange/ TEF colorless |
17.60 €
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
10760903
|
Macherey-Nagel™
70231.2 |
1 mm | Red | Natural rubber / Butyl red orange/ TEF colorless |
16.50 €
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||
10652034
|
Macherey-Nagel™
70231.3 |
1 mm | Blue | Natural rubber / Butyl red orange/ TEF colorless |
16.50 €
Pack of 100 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
Description
- Ready assembled crimp closures and plain crimp caps N 11
- Septa are assembled, caps are ready-to-use.
- Septa are shown for illustration purposes only, images upside down.
- Packs of 100 closures each