Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
Brand:  Macherey-Nagel™ 70231.2
 View more versions of this product
Product Code. 10760903
  • 16.50 € / Pack of 100
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ready assembled crimp closures and plain crimp caps N 11 
  • Septa are assembled, caps are ready-to-use.
  • Septa are shown for illustration purposes only, images upside down.
  • Packs of 100 closures each
Specifications
Specifications
Red
Natural rubber / Butyl red orange/ TEF colorless
1 mm
Aluminum, Butyl Rubber, Rubber, TEF
100 Pk.
Crimp Cap