Learn More
Macherey-Nagel™ N 11 Aluminum Crimp Caps
Seal tubes securely with a variety of crimp closers. Macherey-Nagel™ N 11 aluminum crimp caps come in a variety of styles and colors with different setpa.
Brand:  Macherey-Nagel™ 702995
 View more versions of this product
Product Code. 12490500
  • 31.70 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ready assembled crimp closures and plain crimp caps N 11 
  • Septa are assembled, caps are ready-to-use.
  • Septa are shown for illustration purposes only, images upside down.
  • Packs of 100 closures each
Specifications
Specifications
8 mm
Aluminum, PTFE, Silicone
1 mm
Silver
PTFE Red /Silicone White/PTFE Red