Learn More
N-(2-Pyridyl)thiourea, 98%, Thermo Scientific Chemicals
45.70 € - 153.00 €
Chemical Identifiers
CAS | 14294-11-2 |
---|---|
Molecular Formula | C6H7N3S |
Molecular Weight (g/mol) | 153.20 |
MDL Number | MFCD00041227 |
InChI Key | SLUHLANJIVXTRQ-UHFFFAOYSA-N |
Synonym | 1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea |
PubChem CID | 1490491 |
IUPAC Name | pyridin-2-ylthiourea |
SMILES | NC(=S)NC1=CC=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397786
|
Thermo Scientific Alfa Aesar
L11465.03 |
1 g |
45.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307796
|
Thermo Scientific Alfa Aesar
L11465.06 |
5 g |
153.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(2-Pyridyl)thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
14294-11-2 | |
153.20 | |
SLUHLANJIVXTRQ-UHFFFAOYSA-N | |
1490491 | |
NC(=S)NC1=CC=CC=N1 |
C6H7N3S | |
MFCD00041227 | |
1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea | |
pyridin-2-ylthiourea |
Specifications
14294-11-2 | |
Odorless | |
MFCD00041227 | |
117849 | |
SLUHLANJIVXTRQ-UHFFFAOYSA-N | |
pyridin-2-ylthiourea | |
1490491 | |
98% |
146°C to 149°C | |
C6H7N3S | |
UN2811 | |
1-pyridin-2-yl thiourea, 2-pyridylthiourea, 1-pyridin-2-ylthiourea, 1-2-pyridyl-2-thiourea, n-2-pyridinyl-thiourea, pyridin-2-yl thiourea, pyridin-2-yl-thiourea, n-pyridin-2-ylthiourea, n-2-pyridinyl thiourea, 2-pyridyl thiourea | |
NC(=S)NC1=CC=CC=N1 | |
153.20 | |
153.21 | |
N-(2-Pyridyl)thiourea |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only