missing translation for 'onlineSavingsMsg'
Learn More

Melford N-Acetyl-D-glucosamine

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford A90510-500.0

 View more versions of this product

Product Code. 30018054

  • 554.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 7512-17-6

Molecular Formula: C8H15NO6

Molecular Weight: 221.21

Specifications

Specifications

7512-17-6
C8H15NO6
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers