missing translation for 'onlineSavingsMsg'
Learn More

Melford N-Acetyl-D-glucosamine

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

52.30 € - 554.00 €

Specifications

CAS 7512-17-6
Molecular Formula C8H15NO6
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018053
View Documents
Melford
A90510-5.0
5 g
52.30 €
5g
Please to purchase this item. Need a web account? Register with us today!
30018054
View Documents
Melford
A90510-500.0
500 g
554.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
30018052
View Documents
Melford
A90510-25.0
25 g
113.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 7512-17-6

Molecular Formula: C8H15NO6

Molecular Weight: 221.21

Specifications

Specifications

7512-17-6
C8H15NO6
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.