missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99% Product Code.: 11428770

Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%

Product Code. 11428770
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11428770

Brand: Thermo Scientific Chemicals B21866.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-47-5
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00008681
InChI Key XUYPXLNMDZIRQH-UHFFFAOYNA-N
Synonym n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionineShow MoreShow Less
PubChem CID 6180
SMILES CSCCC(NC(C)=O)C(O)=O

Specifications

Melting Point 113°C to 117°C
Beilstein 1725554
Merck Index 14,96
Solubility Information Soluble in water,ethanol,ethyl acetate.
IUPAC Name 2-acetamido-4-(methylsulfanyl)butanoic acid
Quantity 50 g
Formula Weight 191.25
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-methionine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244892 Certificate TypeCertificate of Analysis Date05/12/2022 Catalog NumberB21866.30,B21866.0B,B21866.18
Lot Number10243571 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog NumberB21866.30,B21866.0B,B21866.18
Lot Number10160445 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21866.30,B21866.0B,B21866.18
Lot NumberY17A041 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21866.30,B21866.0B,B21866.18
Lot Number10227831 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21866.30,B21866.0B,B21866.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.