missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98+% Product Code.: 11458386

Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98+%

Product Code. 11458386
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11458386

Brand: Thermo Scientific Chemicals A15409.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Acetyl-L-cysteine (acetylcysteine, CAS # 616-91-1) is a product of the amino acid L-cysteine.

A mucolytic agent for isolation of mycobacteria from sputum.N-Acetyl-L-cysteine is used in the liquefaction and decontamination of sputum during the preparation for recovery of mycobacterium, which is called as Petroff's method. It shows antiviral activity against the influenza A viruses. It is used in the treatment of interstitial lung disease to prevent disease progression. It is an antioxidant, dietary supplement and protects liver. Further, it forms complexes with palladium, thereby it dissolves in water and remove palladium from drugs. Further, it is used in the treatment of paracetamol overdose. In addition to this, it is involved in the treatment of autism, Alzheimer's disease, bipolar disorder, major depressive disorder, obsessive-compulsive disorder, schizophrenia, and specific drug addictions like cocaine and cannabis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • N-Acetyl-L-cysteine (acetylcysteine) is a precursor of the antioxidant glutathione, or GSH
  • N-Acetyl-L-cysteine is cytoprotective and may prevent cancer, neurodegeneration, and other disease

Application

  • N-Acetyl-L-cysteine is useful for cellular redox study because of its anti-inflammatory properties
  • It may also be useful for psychiatric research and the study of neurodegeneration
  • It is suitable for in vivo and in vitro experiments
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowitShow MoreShow Less
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Melting Point 108°C to 111°C
Beilstein 1724426
Merck Index 14,88
Solubility Information Soluble in water,ethanol,methanol,dimethyl sulfoxide,hot isopropyl alcohol,methyl acetate and ethyl acetate. Insoluble in chloroform and ether.
Quantity 500 g
Formula Weight 163.2
Optical Rotation +5.3° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material N-Acetyl-L-cysteine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM27L020 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA15409.36,A15409.14,A15409.22
Lot NumberZ17K013 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberA15409.14,A15409.36,A15409.22
Lot NumberM20L026 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog NumberA15409.14,A15409.36,A15409.22
Lot Number10252847 Certificate TypeCertificate of Analysis Date28/10/2024 Catalog NumberA15409.14,A15409.36,A15409.22
Lot NumberW06K031 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberA15409.14,A15409.36,A15409.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.