missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyltropinone, 97%, Thermo Scientific™

37.00 € - 362.00 €

Chemical Identifiers

CAS 28957-72-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.30
MDL Number MFCD00151640
InChI Key RSUHKGOVXMXCND-UHFFFAOYNA-N
Synonym 8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
PubChem CID 119846
IUPAC Name 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
SMILES O=C1CC2CCC(C1)N2CC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10081524
View Documents
Thermo Scientific Maybridge
MO07738DA
1 g
37.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10159152
View Documents
Thermo Scientific Maybridge
MO07738EA
10 g
229.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10595544
View Documents
Thermo Scientific Maybridge
MO07738EB
25 g
362.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

28957-72-4
215.30
RSUHKGOVXMXCND-UHFFFAOYNA-N
119846
O=C1CC2CCC(C1)N2CC1=CC=CC=C1
C14H17NO
MFCD00151640
8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
8-benzyl-8-azabicyclo[3.2.1]octan-3-one

Specifications

28957-72-4
C14H17NO
8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
O=C1CC2CCC(C1)N2CC1=CC=CC=C1
215.30
215.29
8-Benzyl-8-azabicyclo[3.2.1]octan-3-one
97%
MFCD00151640
RSUHKGOVXMXCND-UHFFFAOYNA-N
8-benzyl-8-azabicyclo[3.2.1]octan-3-one
119846
97%
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.