Learn More
N-Boc-L-alaninamide, 96%, Thermo Scientific Chemicals
63.90 € - 760.00 €
Chemical Identifiers
CAS | 85642-13-3 |
---|---|
Molecular Formula | C8H16N2O3 |
Molecular Weight (g/mol) | 188.23 |
MDL Number | MFCD03701447 |
InChI Key | GZKKSKKIABUUCX-UHFFFAOYNA-N |
Synonym | boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 |
PubChem CID | 7015295 |
SMILES | CC(NC(=O)OC(C)(C)C)C(N)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419118
|
Thermo Scientific Alfa Aesar
H62604.MD |
250 mg | N/A | ||||||
15499108
|
Thermo Scientific Alfa Aesar
H62604.03 |
1 g |
193.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409118
|
Thermo Scientific Alfa Aesar
H62604.06 |
5 g |
760.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Boc-L-alaninamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-alaninamide is used as pharmaceutical intermediate.
Solubility
Soluble in water or 1% acetic acid.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
85642-13-3 | |
188.23 | |
GZKKSKKIABUUCX-UHFFFAOYNA-N | |
7015295 |
C8H16N2O3 | |
MFCD03701447 | |
boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 | |
CC(NC(=O)OC(C)(C)C)C(N)=O |
Specifications
85642-13-3 | |
MFCD03701447 | |
Soluble in water or 1% acetic acid. | |
CC(NC(=O)OC(C)(C)C)C(N)=O | |
188.23 | |
188.22 | |
Solid |
C8H16N2O3 | |
boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 | |
GZKKSKKIABUUCX-UHFFFAOYNA-N | |
tert-butyl N-(1-carbamoylethyl)carbamate | |
7015295 | |
96% | |
N-Boc-L-alaninamide |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.