missing translation for 'onlineSavingsMsg'
Learn More
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10325592

n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10325592 Shop All Thermo Scientific Products
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
500 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10325592

Brand: Thermo Scientific 181278000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-72-8, 110-54-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
SMILES [Li]CCCC

Specifications

CAS Min % 14.75, 84.75
CAS Max % 15.25, 85.25
Melting Point -95.0°C
Color Pink to White
Density 0.6800g/mL
Boiling Point 60.0°C to 80.0°C
Flash Point −18°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)3Li
Quantity 800 mL
Specific Gravity 0.68
IUPAC Name butyllithium
Formula Weight 64.06
Percent Purity 14.0 to 16.0% (as BuLi) (Active base)
Physical Form Powder or Flakes
Chemical Name or Material n-Butyllithium, 1.6M solution in hexanes
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464899 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number181271000,181275000,181278000,181270100
Lot NumberA0450025 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number181271000,181275000,181278000,181270100
Lot NumberA0451077 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number181271000,181275000,181278000
Lot NumberA0458699 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number181271000,181275000,181278000,181270100
Lot NumberA0435185 Certificate TypeCertificate of Analysis Date31/01/2023 Catalog Number181271000,181275000,181278000,181270100
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butyllithium, 1.6M solution in hexanes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Pyrophoric solid Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H314-Causes severe skin burns and eye damage.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H361f-Suspected of damaging fertility.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Hexane, Butyl lithium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.