Learn More
n-Butyltin hydroxide oxide, 95%, Thermo Scientific Chemicals
66.90 € - 678.00 €
Chemical Identifiers
CAS | 2273-43-0 |
---|---|
Molecular Formula | C4H10O2Sn |
Molecular Weight (g/mol) | 208.83 |
MDL Number | MFCD00013927 |
InChI Key | WIHMDCQAEONXND-UHFFFAOYSA-M |
Synonym | stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid |
PubChem CID | 16767 |
SMILES | CCCC[Sn](O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443837
|
Thermo Scientific Alfa Aesar
A18290.22 |
100 g |
66.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453837
|
Thermo Scientific Alfa Aesar
A18290.36 |
500 g |
220.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463837
|
Thermo Scientific Alfa Aesar
A18290.0E |
2500 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2273-43-0 | |
208.83 | |
WIHMDCQAEONXND-UHFFFAOYSA-M | |
16767 |
C4H10O2Sn | |
MFCD00013927 | |
stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid | |
CCCC[Sn](O)=O |
Specifications
2273-43-0 | |
MFCD00013927 | |
Hygroscopic | |
WIHMDCQAEONXND-UHFFFAOYSA-M | |
butyl(hydroxy)stannanone | |
16767 | |
95% |
C4H10O2Sn | |
UN3146 | |
stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid | |
CCCC[Sn](O)=O | |
208.83 | |
208.81 | |
n-Butyltin hydroxide oxide |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 218-880-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only