missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99+%, extra pure, Thermo Scientific Chemicals

64.62 € - 640.19 €

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10713544
SDS View Documents
Thermo Scientific Acros
383790010
1 L
64.62 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10444263
SDS View Documents
Thermo Scientific Acros
383790025
2.5 L
128.64 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10388402
SDS View Documents
Thermo Scientific Acros
383790050
5 L
228.14 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10227273
SDS View Documents
Thermo Scientific Acros
383790250
25 L
640.19 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC

Specifications

-91.0°C
Colorless to Yellow
0.6830g/mL
−4°C
Glass Bottle
1.3850 to 1.3890
20ppm max.
C7H16
MFCD00009544
0.05% max.
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
Extra Pure
142-82-5
0.003 meq/g max.
98.0°C
Authentic
Liquid
99+%
0.683
CH3(CH2)5CH3
0.4 mPa.s (20°C)
15, 4694
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
CCCCCCC
100.21
CHEBI:43098
≥99%
n-Heptane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

Product Identifier
  • n-Heptane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.