Learn More
n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
54.57 € - 108.54 €
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465065
|
Thermo Scientific Alfa Aesar
047104.AK |
250 mL |
54.57 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475065
|
Thermo Scientific Alfa Aesar
047104.K2 |
1 L |
108.54 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Hexane is used as a solvent, paint thinner, and chemical reaction medium. It is used to extract edible oils from seeds and vegetables, as a special-use solvent, and as a cleaning agent. Hexanes are commonly used in chromatography as a non-polar solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
69°C | |
ChemSeal™ Bottle | |
1.375 | |
CH3(CH2)4CH3 | |
UN1208 | |
14,4694 | |
Insoluble in water | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
n-Hexane, Anhydrous, over molecular sieves |
110-54-3 | |
0.659 g/mL | |
-23°C (-9°F) | |
Liquid | |
C6H14 | |
MFCD02179311 | |
1730733 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: HEXANES
missing translation for 'einecsNumber' : 203-777-6
missing translation for 'rtecsNumber' : MN9275000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
- n-Hexane, anhydrous, over molecular sieves
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Hexane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.