missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Environmental Grade, 95+%, Thermo Scientific Chemicals

For Pesticide Residue, Environmental and GC Analysis

POA - 182.91 €

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359747
View Documents
Thermo Scientific Alfa Aesar
042100.K2
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11379747
View Documents
Thermo Scientific Alfa Aesar
042100.K2
1 L
53.87 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15401787
View Documents
Thermo Scientific Alfa Aesar
042100.M1
4 x 1 L
182.91 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11369747
View Documents
Thermo Scientific Alfa Aesar
042100.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11389747
View Documents
Thermo Scientific Alfa Aesar
042100.M6
4 x 4 L
POA
16 L
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Pesticide residue, environmental and GC analysis.n-Hexane is used in spectrophotometry and environmental testing. It is used as a solvent to extract edible oils from seed and vegetable crops. Commercial grades of hexane are used as solvents for glues, varnishes, and inks, cleaning agent (degreaser) in the printing industry, liquid in low temperature thermometers. other uses are as Fuels and fuel additives, Intermediates and Laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

°C to 95°C
Colorless
0.659
-23°C (-9F)
Weak, characteristic
1.375
C6H14
1730733
Very soluble in ethanol,ethyl ether and chloroform.
CCCCCC
86.18
CHEBI:29021
Environmental
110-54-3
Colorless liquid
69°C
240°C (464°F)
Liquid
>95%
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
n-Hexane
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

missing translation for 'dotInformation' : DOT Class: 3, Packaging Group: II

missing translation for 'einecsNumber' : 203-777-6

missing translation for 'rtecsNumber' : MN9275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • n-Hexane, Environmental Grade

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H361f-Suspected of damaging fertility.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.