missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine, 97%, Thermo Scientific™

58.00 € - 150.00 €

Chemical Identifiers

CAS 879896-58-9
Molecular Formula C12H19N3
Molecular Weight (g/mol) 205.305
MDL Number MFCD09025859
InChI Key CPEUGLUNQVAGDV-UHFFFAOYSA-N
Synonym n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
PubChem CID 17851834
IUPAC Name N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine
SMILES CNCC1=CC(=NC=C1)N2CCCCC2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10517474
View Documents
Thermo Scientific Maybridge
CC50346CB
250 mg Amber Glass Bottle
58.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10326954
View Documents
Thermo Scientific Maybridge
CC50346DA
1 g Amber Glass Bottle
150.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

879896-58-9
205.305
CPEUGLUNQVAGDV-UHFFFAOYSA-N
17851834
CNCC1=CC(=NC=C1)N2CCCCC2
C12H19N3
MFCD09025859
n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine

Specifications

879896-58-9
100.0
C12H19N3
n-methyl-n-2-piperidin-1-ylpyridin-4-yl methyl amine, methyl 2-piperidin-1-yl pyridin-4-yl methyl amine, 4-pyridinemethanamine,n-methyl-2-1-piperidinyl, n-methyl-1-2-piperidin-1-yl pyridin-4-yl methanamine, methyl 2-piperidyl 4-pyridyl methyl amine
CNCC1=CC(=NC=C1)N2CCCCC2
205.305
205.3
N-Methyl-N-[(2-piperidin-1-ylpyridin-4-yl)methyl]amine
96.5
97%
MFCD09025859
CPEUGLUNQVAGDV-UHFFFAOYSA-N
N-methyl-1-(2-piperidin-1-ylpyridin-4-yl)methanamine
17851834
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.