Learn More
N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals
146.00 € - 608.00 €
Chemical Identifiers
CAS | 3197-06-6 |
---|---|
Molecular Formula | C6H16N2O2 |
Molecular Weight (g/mol) | 148.21 |
MDL Number | MFCD00014822 |
InChI Key | CYOIAXUAIXVWMU-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348347
|
Thermo Scientific Alfa Aesar
032312.03 |
1 g |
146.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338347
|
Thermo Scientific Alfa Aesar
032312.06 |
5 g |
608.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3197-06-6 | |
148.21 | |
CYOIAXUAIXVWMU-UHFFFAOYSA-N |
C6H16N2O2 | |
MFCD00014822 |
Specifications
3197-06-6 | |
174°C to 178°C (3 to 4 mmHg) | |
C6H16N2O2 | |
UN2735 | |
Soluble in water. | |
C(CN(CCO)CCO)N | |
148.21 | |
148.21 | |
N,N-Bis(2-hydroxyethyl)ethylenediamine |
1.069 g/mL | |
Amine-like | |
MFCD00014822 | |
n,n-bis 2-hydroxyethyl ethylenediamine, 2,2'-2-aminoethyl azanediyl diethanol, ethanol, 2,2'-2-aminoethyl imino bis, 2,2'-2-aminoethyl imino bisethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethanol, 2-2-aminoethyl 2-hydroxyethyl amino ethan-1-ol, aminoethyldiethanolamine, aminoethylethanolamine solution, 2,2'-2-aminoethyl imino diethanol, acmc-20a9el | |
CYOIAXUAIXVWMU-UHFFFAOYSA-N | |
2-[2-aminoethyl(2-hydroxyethyl)amino]ethanol | |
98535 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only