Learn More
N,N-Dimethyl-1,4-butanediamine, 98%, Thermo Scientific Chemicals
89.14 € - 322.61 €
Chemical Identifiers
CAS | 3529-10-0 |
---|---|
Molecular Formula | C6H16N2 |
Molecular Weight (g/mol) | 116.208 |
MDL Number | MFCD00047977 |
InChI Key | GCOWZPRIMFGIDQ-UHFFFAOYSA-N |
Synonym | 4-dimethylaminobutylamine, 4-aminobutyl dimethylamine, 4-dimethylamino butylamine, n,n-dimethyl-1,4-butanediamine, n,n-dimethylbutane-1,4-diamine, n,n-dimethylaminobutylamine, chembl59625, nn-dimethyl-1,4-diaminobutane, 1,4-butanediamine, n,n-dimethyl, n1,n1-dimethyl-1,4-butanediamine |
PubChem CID | 77063 |
IUPAC Name | N',N'-dimethylbutane-1,4-diamine |
SMILES | CN(C)CCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439410
|
Thermo Scientific Alfa Aesar
H50276.03 |
1 g |
89.14 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449410
|
Thermo Scientific Alfa Aesar
H50276.06 |
5 g |
322.61 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.
Notes
Air sensitive. Store away from oxidizing agents.
Chemical Identifiers
3529-10-0 | |
116.208 | |
GCOWZPRIMFGIDQ-UHFFFAOYSA-N | |
77063 | |
CN(C)CCCCN |
C6H16N2 | |
MFCD00047977 | |
4-dimethylaminobutylamine, 4-aminobutyl dimethylamine, 4-dimethylamino butylamine, n,n-dimethyl-1,4-butanediamine, n,n-dimethylbutane-1,4-diamine, n,n-dimethylaminobutylamine, chembl59625, nn-dimethyl-1,4-diaminobutane, 1,4-butanediamine, n,n-dimethyl, n1,n1-dimethyl-1,4-butanediamine | |
N',N'-dimethylbutane-1,4-diamine |
Specifications
3529-10-0 | |
(CH3)2NCH2CH2CH2CH2NH2 | |
MFCD00047977 | |
Air Sensitive | |
GCOWZPRIMFGIDQ-UHFFFAOYSA-N | |
N',N'-dimethylbutane-1,4-diamine | |
77063 | |
98% |
C6H16N2 | |
1.44 | |
UN2735 | |
4-dimethylaminobutylamine, 4-aminobutyl dimethylamine, 4-dimethylamino butylamine, n,n-dimethyl-1,4-butanediamine, n,n-dimethylbutane-1,4-diamine, n,n-dimethylaminobutylamine, chembl59625, nn-dimethyl-1,4-diaminobutane, 1,4-butanediamine, n,n-dimethyl, n1,n1-dimethyl-1,4-butanediamine | |
CN(C)CCCCN | |
116.208 | |
116.21 | |
N,N-Dimethyl-1,4-butanediamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 222-559-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.