missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

37.19 € - 281.40 €

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10699453
View Documents
Thermo Scientific Acros
396351000
100 mL AcroSeal™ Glass Bottle
37.19 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10424275
View Documents
Thermo Scientific Acros
396350010
1 L AcroSeal™ Glass Bottle
219.09 €
1 L
Please to purchase this item. Need a web account? Register with us today!
13127804
View Documents
Thermo Scientific Acros
396350025
2.5 L AcroSeal™ Glass Bottle
281.40 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

127-19-5
87.12
FXHOOIRPVKKKFG-UHFFFAOYSA-N
31374
N,N-dimethylacetamide
C4H9NO
MFCD00008686
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
CHEBI:84254
CN(C)C(C)=O

Specifications

-20.0°C
99.4
100.0
164.0°C to 166.0°C
Authentic
1.4370 to 1.4390
0.005% max.
C4H9NO
MFCD00008686
0.005% max.
Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
CN(C)C(C)=O
87.12
CHEBI:84254
99.5%
N, N-Dimethylacetamide
127-19-5
Colorless
0.9370g/mL
70°C
Liquid
99.5%
0.937
CH3CON(CH3)2
1.02 mPa.s (20°C)
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
FXHOOIRPVKKKFG-UHFFFAOYSA-N
N,N-dimethylacetamide
31374
87.12
Extra Dry
Videos
Safety and Handling

Safety and Handling

GHS P Statement
May damage the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • N,N-Dimethylacetamide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : dimethyl acetamide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.