missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, >99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 L
Specifications
Specifications
Color | Colorless |
Density | 0.94g/cm3 at 20°C |
Quantity | 1 L |
Physical Form | Liquid |
Assay Percent Range | >99% |
Formula Weight | 87.12 |
Chemical Name or Material | N, N-Dimethylacetamide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Safety and Handling


Product Identifier
- N,N-Dimethylacetamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : N,N-Dimethylacetamide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N,N-Dimethylacetamide, >99%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction