Learn More
N-Propylethylenediamine, 99%, Thermo Scientific Chemicals
56.40 € - 213.00 €
Chemical Identifiers
CAS | 111-39-7 |
---|---|
Molecular Formula | C5H14N2 |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00008172 |
InChI Key | CFNHVUGPXZUTRR-UHFFFAOYSA-N |
Synonym | n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine |
PubChem CID | 66073 |
IUPAC Name | N'-propylethane-1,2-diamine |
SMILES | CCCNCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10195850
|
Thermo Scientific Acros
314750050 |
5 g | Glass bottle |
56.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10651833
|
Thermo Scientific Acros
314750250 |
25 g | Glass bottle |
213.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-39-7 | |
102.18 | |
CFNHVUGPXZUTRR-UHFFFAOYSA-N | |
66073 | |
CCCNCCN |
C5H14N2 | |
MFCD00008172 | |
n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine | |
N'-propylethane-1,2-diamine |
Specifications
111-39-7 | |
100.0 | |
0.8200g/mL | |
14°C | |
99% | |
CH3CH2CH2NHCH2CH2NH2 | |
MFCD00008172 | |
n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine | |
CCCNCCN | |
102.18 | |
102.18 | |
Liquid |
98.5 | |
Colorless | |
147°C to 150°C | |
Authentic | |
C5H14N2 | |
1.4394 to 1.4414 | |
0.82 | |
CFNHVUGPXZUTRR-UHFFFAOYSA-N | |
N'-propylethane-1,2-diamine | |
66073 | |
99% | |
N-Propylethylenediamine, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-864-9
RUO – Research Use Only