Learn More
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide, 97%, Thermo Scientific Chemicals
38.10 € - 411.00 €
Chemical Identifiers
CAS | 77377-52-7 |
---|---|
Molecular Formula | C9H18F3NOSi |
Molecular Weight (g/mol) | 241.329 |
MDL Number | MFCD00009671 |
InChI Key | QRKUHYFDBWGLHJ-UHFFFAOYSA-N |
Synonym | mtbstfa, n-tert-butyldimethylsilyl-n-methyltrifluoroacetamide, n-methyl-n-tert-butyldimethylsilyl trifluoroacetamide, n-tert-butyldimethylsilyl-n-methyl-trifluoroacetamide, n-methyl-n-t-butyldimethylsilyl trifluoroacetamide, acetamide, n-1,1-dimethylethyl dimethylsilyl-2,2,2-trifluoro-n-methyl, n-t-butyldimethylsilyl-n-methyltrifluoroacetamide, n-t-butyldimethylsilyl-n-methyltrifluoro-acetamide |
PubChem CID | 2724275 |
ChEBI | CHEBI:85060 |
IUPAC Name | N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide |
SMILES | CC(C)(C)[Si](C)(C)N(C)C(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418780
|
Thermo Scientific Alfa Aesar
B23760.03 |
1 g |
38.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410208
|
Thermo Scientific Alfa Aesar
B23760.06 |
5 g |
115.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420208
|
Thermo Scientific Alfa Aesar
B23760.14 |
25 g |
411.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It finds its application in the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from hydrolysis of nitrogen mustards and a silylating agent used in the derivatization of various organic compounds (such as amino acids) for gas chromatography-mass spectrometry (GC-MS) analysis and selective O-silylation of N,O-diacylhydroxylamines and GLC derivatization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from hydrolysis of nitrogen mustards and a silylating agent used in the derivatization of various organic compounds (such as amino acids) for gas chromatography-mass spectrometry (GC-MS) analysis and selective O-silylation of N,O-diacylhydroxylamines and GLC derivatization.
Solubility
Soluble in water (reacts).
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store away from electrostatic charges. Keep ignition sources away.
Chemical Identifiers
77377-52-7 | |
241.329 | |
QRKUHYFDBWGLHJ-UHFFFAOYSA-N | |
2724275 | |
N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide |
C9H18F3NOSi | |
MFCD00009671 | |
mtbstfa, n-tert-butyldimethylsilyl-n-methyltrifluoroacetamide, n-methyl-n-tert-butyldimethylsilyl trifluoroacetamide, n-tert-butyldimethylsilyl-n-methyl-trifluoroacetamide, n-methyl-n-t-butyldimethylsilyl trifluoroacetamide, acetamide, n-1,1-dimethylethyl dimethylsilyl-2,2,2-trifluoro-n-methyl, n-t-butyldimethylsilyl-n-methyltrifluoroacetamide, n-t-butyldimethylsilyl-n-methyltrifluoro-acetamide | |
CHEBI:85060 | |
CC(C)(C)[Si](C)(C)N(C)C(=O)C(F)(F)F |
Specifications
77377-52-7 | |
168°C to 170°C | |
C9H18F3NOSi | |
MFCD00009671 | |
3606546 | |
mtbstfa, n-tert-butyldimethylsilyl-n-methyltrifluoroacetamide, n-methyl-n-tert-butyldimethylsilyl trifluoroacetamide, n-tert-butyldimethylsilyl-n-methyl-trifluoroacetamide, n-methyl-n-t-butyldimethylsilyl trifluoroacetamide, acetamide, n-1,1-dimethylethyl dimethylsilyl-2,2,2-trifluoro-n-methyl, n-t-butyldimethylsilyl-n-methyltrifluoroacetamide, n-t-butyldimethylsilyl-n-methyltrifluoro-acetamide | |
QRKUHYFDBWGLHJ-UHFFFAOYSA-N | |
N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide | |
2724275 | |
241.33 | |
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide |
1.023 | |
52°C (125°F) | |
1.402 | |
UN1993 | |
Moisture sensitive | |
Soluble in water (reacts). | |
CC(C)(C)[Si](C)(C)N(C)C(=O)C(F)(F)F | |
241.329 | |
CHEBI:85060 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only