missing translation for 'onlineSavingsMsg'
Learn More

n-Undecanoyl chloride, 98%, Thermo Scientific Chemicals

112.00 € - 198.00 €

Chemical Identifiers

CAS 17746-05-3
Molecular Formula C11H21ClO
Molecular Weight (g/mol) 204.738
MDL Number MFCD00000739
InChI Key JUKPJGZUFHCZQI-UHFFFAOYSA-N
Synonym n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
PubChem CID 87287
IUPAC Name undecanoyl chloride
SMILES CCCCCCCCCCC(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442848
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H56380.ME
500 mg
112.00 €
500mg
Please to purchase this item. Need a web account? Register with us today!
15432848
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H56380.03
1 g
198.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17746-05-3
204.738
JUKPJGZUFHCZQI-UHFFFAOYSA-N
87287
CCCCCCCCCCC(=O)Cl
C11H21ClO
MFCD00000739
n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
undecanoyl chloride

Specifications

17746-05-3
113°C (235°F)
CH3(CH2)9COCl
MFCD00000739
1759226
n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
CCCCCCCCCCC(=O)Cl
204.738
204.74
n-Undecanoyl chloride
135°C to 136°C
C11H21ClO
1.443
UN3265
Moisture sensitive
JUKPJGZUFHCZQI-UHFFFAOYSA-N
undecanoyl chloride
87287
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 241-741-1

missing translation for 'tsca' : Yes

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only