Learn More
N4-Acetylcytidine triphosphate sodium, MedChemExpress
MedChemExpress N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro transcription and it can be incorporated into multiple templates.
Brand:  Medchem Express HY-111815A
Encompass_Preferred
 View more versions of this product
Product Code. 16434538
  • 1674.00 € / 10mg
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro
Specifications
Specifications
N4-Acetylcytidine triphosphate sodium
Research
10 mg
H2O : ≥ 250 mg/mL (407.75 mM)
613.12
Research
Off-White
C11H14N3Na4O15P3
ac4CTP sodium
O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)NC(C)=O)=O)O2)O)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O
613.12
Solid