Learn More
N4-Acetylcytidine triphosphate sodium, MedChemExpress
MedChemExpress N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitro transcription and it can be incorporated into multiple templates.
1048.95 € - 9031.50 €
Specifications
Chemical Name or Material | N4-Acetylcytidine triphosphate sodium |
---|---|
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C11H14N3Na4O15P3 |
Synonym | ac4CTP sodium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16495386
|
Medchem Express
HY-111815A |
100 mg |
9031.50 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16477247
|
Medchem Express
HY-111815A |
5 mg |
1048.95 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16470837
|
Medchem Express
HY-111815A |
50 mg |
5644.35 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434538
|
Medchem Express
HY-111815A |
10 mg |
1674.00 €
10mg |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
N4-Acetylcytidine triphosphate sodium is efficiently used as a substrate in T7 Polymerase-catalyzed in vitroSpecifications
N4-Acetylcytidine triphosphate sodium | |
Research | |
ac4CTP sodium | |
O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)NC(C)=O)=O)O2)O)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O | |
613.12 | |
Solid |
Off-White | |
C11H14N3Na4O15P3 | |
H2O : ≥ 250 mg/mL (407.75 mM) | |
613.12 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.