missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Nailsaver Sponges
Unique side grooves protect the fingernails, while also allowing for a better grip for more cleaning leverage. This pad protects fingernails for worry free washing up, reducing the need for gloves and the likelihood of broken nails.
Brand: 3M™ 7000097765
This item is not returnable.
View return policy
Description
- Cleaning sponges with a unique ridge to protect finger nails
- Handy size for a good grip and excellent durability
- Foam shape is easy to handle
- Reduces the need for gloves
- Extremely long lasting

Specifications
Sponge | |
92 x 70 mm | |
150 mm | |
10 Pk. | |
70 mm |
Pink | |
Cleaning | |
Foam | |
Nailsaver |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction