missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.

Brand:  Thermo Scientific™ 2650-0020

814.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10050461

  • 836.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.

Features:

  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction

Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Comply with FDA Reg. 177.1520 and USP Class VI
47 cm
Polypropylene
75 L
81 cm
18.5 in.
Closed-dome Tank
20 gal.
35 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks > Capacity: 20 gal. (75L)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.