Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
Brand:  Thermo Scientific™ 2650-0030
 View more versions of this product
Product Code. 10338990
  • 1185.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Comply with FDA Reg. 177.1520 and USP Class VI
47 cm
Polypropylene
115 L
98.1 cm
18.5 in.
Closed-dome Tank
30 gal.
38.625 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.