missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers
GREENER_CHOICE

Store and transport dry and solid materials, specimens and general lab supplies in these stackable containers which feature positive-lock lids that form a tight seal.

Brand:  Thermo Scientific™ 7142-0015

 View more versions of this product

Product Code. 12726737

  • 536.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Store and transport dry and solid materials, specimens and general lab supplies using Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers. Containers are stackable and have positive-lock lids that seal tight for storing both dry and solid materials.

Features:
  • High-density, polyethylene containers provide excellent chemical and temperature resistance
  • Easy-grip, molded-in handles are comfortable and durable for lifting and moving
  • Positive-lock lid provides a tight seal for transport or storage
  • NSF approved for food and beverage storage
TRUSTED_SUSTAINABILITY
Spécification

Spécification

Container with Cover
4 gal.
NSF approved for food and beverage storage.
30.5 cm
9 in.
Lid
HDPE
15 L
Container Large Round w/Cover HDPE 15 L
Molded
24 cm
1 Ea.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Product Certifications
Promotions

Promotions