Learn More
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers
GREENER_CHOICE
Store and transport dry and solid materials, specimens and general lab supplies in these stackable containers which feature positive-lock lids that form a tight seal.
61.56 € - 557.44 €
Specifications
Material HDPE
Certifications/Compliance NSF approved for food and beverage storage.
Graduations Molded
Includes Lid
No. per Pack 1 Ea.
View More Specs
Products 3
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
12726737
GREENER_CHOICE
View Documents
Thermo Scientific™
7142-0015
15 L 30.5 cm 24 cm
557.44 €
Pack of 6
10070631
GREENER_CHOICE
View Documents
Thermo Scientific™
7142-0015
15 L 30.5 cm 24 cm N/A N/A N/A
10204311
GREENER_CHOICE
View Documents
Thermo Scientific™
7142-0020
20 L 30.5 cm 38 cm
112.00 €
Each
Description
Description
Store and transport dry and solid materials, specimens and general lab supplies using Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers. Containers are stackable and have positive-lock lids that seal tight for storing both dry and solid materials.
Features:
  • High-density, polyethylene containers provide excellent chemical and temperature resistance
  • Easy-grip, molded-in handles are comfortable and durable for lifting and moving
  • Positive-lock lid provides a tight seal for transport or storage
  • National Sanitation Foundation (NSF) approved for food and beverage storage
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Specifications
HDPE
Molded
1 Ea.
NSF approved for food and beverage storage.
Lid
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers