Learn More
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles

Description
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Polypropylene screw closure/stem with PPCO draw tube
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 32 oz., 1000 mL |
Capacity (Metric) | 1000 mL |
Capacity (English) | 32 oz. |
Color | Translucent |
Includes | Stem and draw tube |
Material | LDPE |
No. per Case | 12 |
No. per Pack | 4 |
Closure Included | Yes |
Graduated | No |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7452386010 | Certificate of Compliance | 31/03/2025 | 2401-1000 |
7436485010 | Certificate of Compliance | 13/01/2025 | 2401-1000 |
7425265010 | Certificate of Compliance | 13/01/2025 | 2401-1000 |
7313933010 | Certificate of Compliance | 25/07/2020 | 2401-1000 |
7273013010 | Certificate of Compliance | 20/12/2017 | 2401-1000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.