Learn More
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles
GREENER_CHOICE
Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
23.40 € - 165.00 €
Specifications
Color Translucent
Includes Stem and draw tube
Material LDPE
Closure Included Yes
Graduated No
View More Specs
Products 4
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10709121
GREENER_CHOICE
View Documents
Thermo Scientific™
2401-0125
125 mL 24 mm
23.40 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
10624301
GREENER_CHOICE
View Documents
Thermo Scientific™
2401-0250
250 mL 24 mm
34.20 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
11973857
GREENER_CHOICE
View Documents
Thermo Scientific™
2401-0500
500 mL 28 mm
165.00 €
Pack of 24
In Stock
to see stock.
Edge
Add to basket
10048300
GREENER_CHOICE
View Documents
Thermo Scientific™
2401-1000
1000 mL 38-430 mm
28.76 €
Pack of 4
In Stock
to see stock.
Edge
Add to basket
Description
Description
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:
  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Specifications
Translucent
LDPE
No
Cylindrical
Polypropylene
Yes
Stem and draw tube
Yes
Narrow
Economy Wash Bottle
Screw Cap
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles