missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure

Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.

45.63 € - 1054.00 €

Specifications

Color Translucent
Description Narrow Mouth Bottle
For Use With (Application) Storage, Transportation ​
Material Teflon PFA
No. per Pack 1 Ea.
View More Specs

Products 6
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10414671
View Documents Promotion Details
Thermo Scientific™
DS1630-0001
30 mL 20 mm
45.63 €
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15911661
View Documents Promotion Details
Thermo Scientific™
DS1630-0002
60 mL 20 mm
437.00 €
Pack of 8
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15911381
View Documents Promotion Details
Thermo Scientific™
1630-0004
125 mL 38-430 mm
510.00 €
Pack of 6
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11329774
View Documents Promotion Details
Thermo Scientific™
1630-0008
250 mL 38-430 mm
484.00 €
Pack of 4
View alternative products
Save Up to 
»
Estimated Shipment: 04-11-2024
to see stock.
Add to basket
 
10758331
View Documents Promotion Details
Thermo Scientific™
1630-0016
500 mL 38-430 mm
166.25 €
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11349774
View Documents Promotion Details
Thermo Scientific™
1630-0032
1 L 38-430 mm
1054.00 €
Pack of 4
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

Get excellent resistance to temperature, chemicals and corrosion with Thermo Scientific™ Nalgene™ Narrow-Mouth Bottles Made of Teflon™ PFA with Closure, among the most chemical- and corrosion-resistant containers available.

  • Withstand -270°C to +250°C (-454°F to +482°F)
  • Inert to virtually all chemicals except molten alkali metals, fluorine at high temperatures and complex halogenated compounds at high temperatures and pressures
  • Packaged individually
  • Autoclavable
  • Leakproof† PFA closure

Includes:

Leakproof, linerless PFA screw caps

Note:

Completely disengage threads or remove cap before autoclaving

Warranty:

90 days

Specifications

Specifications

Translucent
Storage, Transportation ​
1 Ea.
-100°C to 260°C
Yes
Narrow
Narrow Mouth Bottle
ETFE
No
Narrow Mouth Bottle
Teflon PFA
Non-sterile
Natural
Autoclavable
Round
Unlined
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.