Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE
Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
10.20 € - 113.00 €
Specifications
Closure Included Yes
View More Specs
Products 14
Product Code Brand Product Type Closure Size Color Price Quantity & Availability  
Product Code Brand Product Type Closure Size Color Price Quantity & Availability  
10586551
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712151-0200
Replacement Cap 20 mm White
10.20 €
Pack of 12
In Stock
to see stock.
Add to basket
10264401
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712150-0240
Replacement Cap 24 mm Natural
10.90 €
Pack of 12
In Stock
to see stock.
Add to basket
10020661
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712150-0280
Replacement Cap 28 mm Natural
11.50 €
Pack of 12
In Stock
to see stock.
Add to basket
11334595
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712172-0020
Stem and Draw Tube 28 mm Natural
25.00 €
Pack of 2
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
10010681
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712160-0384
Replacement Cap 38 mm Natural
22.20 €
Pack of 12
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
10603452
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712151-0380
Replacement Cap 38 mm White
13.15 €
Pack of 12
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
10799362
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712150-0380
Screw Cap 38 mm Natural
14.10 €
Pack of 12
In Stock
to see stock.
Add to basket
10274401
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712150-0430
Replacement Cap 43 mm Natural
13.65 €
Pack of 12
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
10162491
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
712171-0530
Replacement Cap 53 mm Amber
22.85 €
Pack of 12
In Stock
to see stock.
Add to basket
10000721
GREENER_CHOICE
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific™
712174-0530
Replacement Cap 53 mm Natural
41.05 €
Pack of 2
Estimated Shipment: 07-04-2025
to see stock.
Zum Warenkorb hinzufügen
10050681
GREENER_CHOICE
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific™
712150-0530
Replacement Cap 53 mm Natural
19.90 €
Pack of 12
In Stock
to see stock.
Zum Warenkorb hinzufügen
10365941
GREENER_CHOICE
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific™
712150-0630
Replacement Cap 63 mm Natural
28.85 €
Pack of 12
Estimated Shipment: 01-04-2025
to see stock.
Zum Warenkorb hinzufügen
12716677
GREENER_CHOICE
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific™
712411-1030
Spout Cap 20 mm Natural
108.00 €
Pack of 25
In Stock
to see stock.
Zum Warenkorb hinzufügen
10644212
GREENER_CHOICE
missing translation for 'viewDocuments' Aktionsdetails
Thermo Scientific™
712411-1125
Spout Cap 24 mm Natural
113.00 €
Pack of 25
In Stock
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.
  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
  • Leakproof† when used with the Nalgene bottles with the matching finish
.
Spezifikation
Spezifikation
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.