missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Certified Clean Biotainer Bottle
For Use With: Specifically designed for pharmaceutical and biological manufacturing; Closure Material: Polypropylene; Color: Clear
728.00 € - 1415.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | USP <778> |
Closure Color | White |
Closure Liner | LDPE |
Closure Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Includes | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Quantity | Price | Quantity & Availability | |||
16410591
|
Thermo Scientific™
383025-42 |
125 mL | Side Handle | 20 x 5 Bottles |
1310.00 €
Pack of 100 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||
16400591
|
Thermo Scientific™
383005-42 |
500 mL | Side Handle | 14 x 5 Bottles |
1415.00 €
Pack of 70 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||
16430591
|
Thermo Scientific™
383110-42 |
1 L | Side Handle | 7 x 5 Bottles |
1405.00 €
Pack of 35 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||
16460591
|
Thermo Scientific™
383230-42 |
2 L | Side Handle | 4 x 5 Bottles |
1225.00 €
Pack of 20 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||
16450591
|
Thermo Scientific™
383230-20 |
2 L | Side Handle | 20 Bottles |
1210.00 €
Pack of 20 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
16420601
|
Thermo Scientific™
383415-16 |
5 L | Side Handle | 6 Bottles |
754.00 €
Pack of 6 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||
16430601
|
Thermo Scientific™
383415-42 |
5 L | Neck Handle | 6 Bottles |
728.00 €
Pack of 6 |
In Stock
Log in to see stock. |
|||
Description
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.
- Specifically designed for pharmaceutical and biological manufacturing
- Certified Clean, validated to USP <788>
- <0.5 EU/mL Endotoxin
Specifications
Not Autoclavable | |
White | |
Polypropylene | |
Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product | |
Biotainer | |
USP <788> (Low particulate) |
USP <778> | |
LDPE | |
Clear | |
PETG | |
Sterile |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title