Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Crimp Finish: Nonsterile, Shrink-Wrapped Modules

Protect against leakage, breakage, and contamination. Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.
Brand: Thermo Scientific™ 322030-0010
632.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- Nonsterile
Ordering Information:
Serum Vials are packed in trayless shrink-wrapped modules. Stoppers and Caps not included.

Specifications
Serum Vials | |
Clear | |
PETG | |
10 mL | |
252 | |
Nalgene | |
Stoppers and Caps (Not Included) | |
252 Pk., 1260 Cases | |
Serum Insert Vial | |
Lined Continuous Thread Closure |
13 mm | |
24 mm | |
Non-sterile | |
1260 | |
1/3 oz. | |
10 mL | |
50 mm | |
ISO 13485:2003 | |
20 mm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1402879 | Certificate of Compliance | 17/03/2024 | 322030-0010 |
1390280 | Certificate of Compliance | 25/08/2023 | 322030-0010 |
1377847 | Certificate of Compliance | 22/02/2023 | 322030-0010 |
1294350 | Certificate of Compliance | 10/09/2020 | 322030-0010 |
1271339 | Certificate of Compliance | 06/02/2020 | 322030-0010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.