Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand:  DWK Life Sciences W987000-S
 View more versions of this product
Product Code. 17922581
  • 1110.00 € / Pack of 278
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
Certifications
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Sterile
278/Cs.
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Borosilicate Glass (Type 1)
2 mL
Wheaton CompletePAK
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials > 2 mL; Clear, 278/Cs.