missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.

Brand:  Thermo Scientific™ 342030-0020

314.68 EUR valid until 2024-11-29
Use promo code "21718" to get your promotional price.


 View more versions of this product

Product Code. 10632824

  • 427.23 € / Pack of 612
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Vials
Clear
PETG
20 mL
153
Nalgene
Stoppers and Caps (Not Included)
612 Cases, 153 Pk.
Serum Vial
Stopper, Cap
13 mm
30 mm
Sterile
612
2/3 oz.
20 mL
58 mm
ISO 13485:2016
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers