missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels
GREENER_CHOICE

Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.

Brand:  Thermo Scientific™ 6319-0020

56.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11319821

  • 56.80 € / Pack of 300
Estimated Shipment: 22-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.

Features:

  • Durable polyolefin labels
  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt, and won't discolor, crack or tear
  • Excellent for labeling microscopic slides—adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents, although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack

Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyolefin Label, 1 x 2.63
Polyolefin
1500/Cs.
2.63 in.
1 in.
Label
30
300/Pk.
67 mm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels > 30 labels/sheet; L x W: 1 x 2.63 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.