Learn More
Thermo Scientific™ Nalgene™ Polypropylene Stirring Rod

Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Brand: Thermo Scientific™ 6169-0010
49.17 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.
- Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
- Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
- Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications

Specifications
Autoclavable | |
Stirring Rod with flat paddle and triangular paddle ends | |
248 mm | |
Stirring Rod |
White | |
9¾ in. | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1423138 | Certificate of Compliance | 08/03/2025 | 6169-0010 |
1413260 | Certificate of Compliance | 11/09/2024 | 6169-0010 |
1397694 | Certificate of Compliance | 03/12/2023 | 6169-0010 |
1389152 | Certificate of Compliance | 08/07/2023 | 6169-0010 |
1376642 | Certificate of Compliance | 16/12/2022 | 6169-0010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.