missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles Product Code.: 10618101
Add to basket

Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles
GREENER_CHOICE

Product Code. 10618101
500 mL, 28 mm
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Size:
28 mm
38-430 mm
This item is not returnable. View return policy

Product Code. 10618101

Brand: Thermo Scientific™ 24050500

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.

Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.

  • Polypropylene screw/stem and draw tube
  • Tip can be cut back to provide increased flow
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Includes Screw/stem and draw tube
Material PPCO
No. per Case 24
No. per Pack 6
Sterility Non-sterile
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 213 mm
Product Type Wash Bottle
Height (English) 8.4 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 72.6 mm
Closure Type Cap
Through the Closure Dispensing Yes
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7429406010 Certificate TypeCertificate of Compliance Date10/03/2025 Catalog Number2405-0500
Lot Number7449223010 Certificate TypeCertificate of Compliance Date22/01/2025 Catalog Number2405-0500
Lot Number7445241010 Certificate TypeCertificate of Compliance Date20/01/2025 Catalog Number2405-0500
Lot Number7420426010 Certificate TypeCertificate of Compliance Date20/12/2024 Catalog Number2405-0500
Lot Number7318029010 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2405-0500
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.