missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge
GREENER_CHOICE

Quickly attain and hold vacuum of 25 in. (635 mm) Hg with these hand-operated vacuum pumps featuring a new, more durable and ergonomic handle design.

Brand:  Thermo Scientific™ 6131-0010

143.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10008570

  • 193.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Quickly attain and hold vacuum of 25 in. (635 mm) Hg with Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge. These pumps feature a new, more durable and ergonomic handle design. This repairable economy version is ideal for field or lab use.

Easy and Convenient Operation

  • Polyvinyl Chloride (PVC) body
  • Ergonomic vacuum trigger release requires only one hand to operate and releases vacuum with the touch of an index finger
  • Adjustable vacuum release rate provides control
  • Full blank port pressure is obtained with only two strokes, making operation efficient
  • Removable cover on exhaust port and trigger to release vacuum without disconnecting pumps from the line
  • Economical design without vacuum gauge 
  • Adjustable vacuum release rate controlled by trigger on pump
  • Nozzle fits standard 0.25″ I.D. tubing
  • Can be disassembled for repair and replacement of internal parts (see catalog numbers for repair kits in the table below)

Includes:

Supplied with 2 ft. length of Clear Plastic Tubing

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Repairable Hand Vacuum Pump
4
15 cc/stroke
6132-2000 Repair Kit
1
25 in. Hg
Vacuum Pump
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge > Repairable Hand Vacuum Pumps; Pumping rate/stroke: 15cc

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.