missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.

Brand:  Thermo Scientific™ 4112-0500

 View more versions of this product

Product Code. 10179661

  • 146.00 € / Pack of 12
Estimated Shipment: 15-09-2026
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features:
  • Sterile and single use: Reduce contamination risk
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
TRUSTED_SUSTAINABILITY
Specifications

Specifications

16 oz.
PETG
1
None
Single-use
Narrow
Plain
45-430 mm
500 mL
12
Erlenmeyer
Not Autoclavable
Yes
Sterile
Solid, non-vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.