Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
75.85 € - 316.00 €
Specifications
Material PETG
No. per Pack 1
Shape Erlenmeyer
Surface Coating None
Autoclavable Not Autoclavable
View More Specs
Products 12
Product Code Brand Capacity (Metric) Closure Size Closure Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Closure Type Price Quantity & Availability  
10266432
View accessory products View Documents
Thermo Scientific™
4115-0125
125 mL 38-430 mm Vented
316.00 €
Pack of 24
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
10592033
View accessory products View Documents
Thermo Scientific™
4115-0250
250 mL 38-430 mm Vented
174.00 €
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10686673
View accessory products View Documents
Thermo Scientific™
4115-0500
500 mL 45-430 mm Vented
212.00 €
Pack of 12
In Stock
to see stock.
Edge
Add to basket
10357302
View accessory products View Documents
Thermo Scientific™
4115-1000
1000 mL 45-430 mm Vented
137.00 €
Pack of 6
In Stock
to see stock.
Add to basket
10473582
View accessory products View Documents
Thermo Scientific™
4115-2000
2000 mL 45-430 mm Vented
89.20 €
Pack of 4
In Stock
to see stock.
Edge
Add to basket
10156192
View Documents
Thermo Scientific™
4115-2800
2800 mL 70 mm Vented
168.00 €
Pack of 4
In Stock
to see stock.
Edge
Add to basket
10465622
View accessory products View Documents
Thermo Scientific™
4112-0125
125 mL 38-430 mm Solid, non-vented
212.00 €
Pack of 24
In Stock
to see stock.
Edge
Add to basket
10292032
View accessory products View Documents
Thermo Scientific™
4112-0250
250 mL 38-430 mm Solid, non-vented
140.00 €
Pack of 12
In Stock
to see stock.
Add to basket
10179661
View accessory products View Documents
Thermo Scientific™
4112-0500
500 mL 45-430 mm Solid, non-vented
146.00 €
Pack of 12
In Stock
to see stock.
Add to basket
10475622
View accessory products View Documents
Thermo Scientific™
4112-1000
1000 mL 45-430 mm Solid, non-vented
75.85 €
Pack of 6
In Stock
to see stock.
Add to basket
10385012
View accessory products View Documents
Thermo Scientific™
4112-2000
2000 mL 45-430 mm Solid, non-vented
81.95 €
Pack of 4
In Stock
to see stock.
Add to basket
10228732
View Documents
Thermo Scientific™
2800 mL 70 mm Solid, non-vented
163.00 €
Pack of 4
In Stock
to see stock.
Add to basket
Description
Description
Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:
  • Sterile and single use: Reduce contamination risk
  • BPA-free
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
  • Regulatory documentation available on request
Warranty: 90 days
Specifications
Specifications
PETG
Erlenmeyer
Not Autoclavable
Yes
Plain
1
None
Single-use
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile